Assay Details
| Gene Symbol: DAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044682.1 | XP_017900171.1 | 3 - 4 | 439 | 58 | |
| XM_018044685.1 | XP_017900174.1 | 3 - 4 | 439 | 58 | ||
More Information
Set Membership: |
|

DAB1
Genomic Map