Assay Details
| Gene Symbol: GZMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682880.3 | XP_005682937.2 | 2 - 3 | 233 | 79 | |
| GenBank mRNA | JO599354.2 | - | 2 - 3 | 290 | 79 | |
More Information
Set Membership: |
|

GZMM
Genomic Map