Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684176.3 | XP_005684233.2 | 7 - 8 | 2342 | 66 | |
| XM_005684179.3 | XP_005684236.2 | 8 - 9 | 2091 | 66 | ||
| XM_018052506.1 | XP_017907995.1 | 6 - 7 | 2204 | 66 | ||
| XM_018052507.1 | XP_017907996.1 | 8 - 9 | 2414 | 66 | ||
More Information
Set Membership: |
|

PTPDC1
Genomic Map