Assay Details
| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686002.3 | XP_005686059.1 | 6 - 7 | 1109 | 73 | |
| XM_013967249.2 | XP_013822703.2 | 7 - 8 | 1264 | 73 | ||
More Information
Set Membership: |
|

RDH11
Genomic Map