Assay Details
| Gene Symbol: SLC20A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055327.1 | XP_017910816.1 | 10 - 11 | 4581 | 57 | |
| GenBank mRNA | JO645765.2 | - | 10 - 11 | 3298 | 57 | |
More Information
Set Membership: |
|

SLC20A1
Genomic Map