Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687975.3 | XP_005688032.1 | 4 - 5 | 1009 | 70 | |
| XM_005687976.3 | XP_005688033.1 | 4 - 5 | 1003 | 70 | ||
| XM_018057151.1 | XP_017912640.1 | 4 - 5 | 1148 | 70 | ||
| GenBank mRNA | JO432577.2 | - | 1 - 2 | 711 | 70 | |
More Information
Set Membership: |
|

PTER
Genomic Map