Assay Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688518.3 | XP_005688575.1 | 3 - 4 | 744 | 66 | |
| XM_018057813.1 | XP_017913302.1 | 3 - 4 | 489 | 66 | ||
More Information
Set Membership: |
|

MYL9
Genomic Map