Assay Details
| Gene Symbol: METTL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693325.3 | XP_005693382.2 | 7 - 8 | 924 | 76 | |
| GenBank mRNA | JT704027.1 | - | 4 - 5 | 911 | 76 | |
More Information
Set Membership: |
|

METTL16
Genomic Map