Assay Details
| Gene Symbol: ELOVL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065758.1 | XP_017921247.1 | 2 - 3 | 235 | 71 | |
| GenBank mRNA | JO434829.2 | - | 2 - 3 | 238 | 71 | |
| KA317595.1 | - | 7 - 8 | 2665 | 71 | ||
More Information
Set Membership: |
|

ELOVL7
Genomic Map