Assay Details
| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696573.3 | XP_005696630.1 | 4 - 5 | 690 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAPCD1| Gene Symbol: SAPCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696573.3 | XP_005696630.1 | 4 - 5 | 690 | 66 | |
Set Membership: |
|