Assay Details
| Gene Symbol: LOC102186759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051323.1 | XP_017906812.1 | 2 - 3 | 312 | 85 | |
| XM_018051324.1 | XP_017906813.1 | 2 - 3 | 324 | 85 | ||
More Information
Set Membership: |
|

LOC102186759
Genomic Map