Assay Details
| Gene Symbol: LOC102186759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051323.1 | XP_017906812.1 | 3 - 4 | 454 | 84 | |
| XM_018051324.1 | XP_017906813.1 | 3 - 4 | 466 | 84 | ||
More Information
Set Membership: |
|

LOC102186759
Genomic Map