Assay Details
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053891.1 | XP_017909380.1 | 6 - 7 | 899 | 64 | |
GenBank mRNA | JO380986.2 | - | 6 - 7 | 820 | 64 | |
JO424891.1 | - | 6 - 7 | 971 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
