Assay Details
| Gene Symbol: PROX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970386.2 | XP_013825840.1 | 3 - 4 | 2459 | 87 | |
| XM_013970387.2 | XP_013825841.1 | 3 - 4 | 2728 | 87 | ||
| XM_013970389.2 | XP_013825843.1 | 3 - 4 | 2040 | 87 | ||
| XM_018060736.1 | XP_017916225.1 | 3 - 4 | 2160 | 87 | ||
| XM_018060737.1 | XP_017916226.1 | 3 - 4 | 2728 | 87 | ||
| GenBank mRNA | JO419520.2 | - | 2 - 3 | 1288 | 87 | |
More Information
Set Membership: |
|

PROX1
Genomic Map