Assay Details
| Gene Symbol: KCTD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061095.1 | XP_017916584.1 | 5 - 6 | 606 | 107 | |
| XM_018061096.1 | XP_017916585.1 | 5 - 6 | 603 | 107 | ||
| GenBank mRNA | JO413989.2 | - | 5 - 6 | 601 | 107 | |
| JO413990.2 | - | 4 - 5 | 656 | 107 | ||
More Information
Set Membership: |
|

KCTD10
Genomic Map