Assay Details
| Gene Symbol: MLKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970972.2 | XP_013826426.1 | 7 - 8 | 1351 | 66 | |
| XM_018061703.1 | XP_017917192.1 | 7 - 8 | 1372 | 66 | ||
| XM_018061704.1 | XP_017917193.1 | 7 - 8 | 1249 | 66 | ||
More Information
Set Membership: |
|

MLKL
Genomic Map