Assay Details
| Gene Symbol: LOC102171556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061126.1 | XP_017916615.1 | 22 - 23 | 2603 | 89 | |
| GenBank mRNA | KA337612.1 | - | 13 - 14 | 1781 | 89 | |
More Information
Set Membership: |
|

LOC102171556
Genomic Map