Assay Details
| Gene Symbol: LOC102171556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061126.1 | XP_017916615.1 | 27 - 28 | 3288 | 63 | |
| GenBank mRNA | KA337612.1 | - | 8 - 9 | 1096 | 63 | |
More Information
Set Membership: |
|

LOC102171556
Genomic Map