Assay Details
Gene Symbol: LOC102171556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061126.1 | XP_017916615.1 | 3 - 4 | 336 | 57 | |
GenBank mRNA | KA305266.1 | - | 2 - 3 | 237 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
