Assay Details
| Gene Symbol: LOC102171556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061126.1 | XP_017916615.1 | 4 - 5 | 445 | 83 | |
| GenBank mRNA | KA305266.1 | - | 1 - 2 | 128 | 83 | |
More Information
Set Membership: |
|

LOC102171556
Genomic Map