Assay Details
| Gene Symbol: LOC102179132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064502.1 | XP_017919991.1 | 10 - 11 | 2884 | 110 | |
| XM_018064503.1 | XP_017919992.1 | 11 - 12 | 2906 | 110 | ||
| GenBank mRNA | JO548077.2 | - | 11 - 12 | 1571 | 110 | |
| KA309795.1 | - | 8 - 9 | 2112 | 110 | ||
More Information
Set Membership: |
|

LOC102179132
Genomic Map