Assay Details
| Gene Symbol: LOC102179132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064502.1 | XP_017919991.1 | 12 - 13 | 3153 | 96 | |
| XM_018064503.1 | XP_017919992.1 | 13 - 14 | 3175 | 96 | ||
| GenBank mRNA | JO548077.2 | - | 13 - 14 | 1840 | 96 | |
| KA309795.1 | - | 10 - 11 | 2381 | 96 | ||
More Information
Set Membership: |
|

LOC102179132
Genomic Map