Assay Details
| Gene Symbol: LOC102179132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064502.1 | XP_017919991.1 | 5 - 6 | 2152 | 104 | |
| XM_018064503.1 | XP_017919992.1 | 6 - 7 | 2174 | 104 | ||
| GenBank mRNA | JO332083.2 | - | 5 - 6 | 894 | 104 | |
| JO548077.2 | - | 6 - 7 | 839 | 104 | ||
| KA309795.1 | - | 6 - 7 | 963 | 104 | ||
More Information
Set Membership: |
|

LOC102179132
Genomic Map