Assay Details
| Gene Symbol: LOC102179132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064502.1 | XP_017919991.1 | 8 - 9 | 2595 | 89 | |
| XM_018064503.1 | XP_017919992.1 | 9 - 10 | 2617 | 89 | ||
| GenBank mRNA | JO332083.2 | - | 2 - 3 | 451 | 89 | |
| JO548077.2 | - | 9 - 10 | 1282 | 89 | ||
More Information
Set Membership: |
|

LOC102179132
Genomic Map