Assay Details
| Gene Symbol: PRKAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972630.2 | XP_013828084.1 | 2 - 3 | 523 | 64 | |
| XM_018063935.1 | XP_017919424.1 | 2 - 3 | 288 | 64 | ||
| GenBank mRNA | JO634298.2 | - | 9 - 10 | 3144 | 64 | |
| JO634299.2 | - | 9 - 10 | 3144 | 64 | ||
More Information
Set Membership: |
|

PRKAR1A
Genomic Map