Assay Details
| Gene Symbol: ARSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063936.1 | XP_017919425.1 | 4 - 5 | 1079 | 54 | |
| XM_018063938.1 | XP_017919427.1 | 3 - 4 | 1021 | 54 | ||
| GenBank mRNA | JO429218.2 | - | 9 - 10 | 1821 | 54 | |
More Information
Set Membership: |
|

ARSG
Genomic Map