Assay Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038731.1 | XP_017894220.1 | 4 - 5 | 1043 | 61 | |
| XM_018038732.1 | XP_017894221.1 | 5 - 6 | 858 | 61 | ||
| GenBank mRNA | JO618937.2 | - | 3 - 4 | 1158 | 61 | |
| KA330379.1 | - | 4 - 5 | 717 | 61 | ||
More Information
Set Membership: |
|

BYSL
Genomic Map