Assay Details
| Gene Symbol: IGSF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054731.1 | XP_017910220.1 | 4 - 5 | 811 | 54 | |
| GenBank mRNA | JO639950.2 | - | 4 - 5 | 467 | 54 | |
More Information
Set Membership: |
|

IGSF21
Genomic Map