Assay Details
| Gene Symbol: PADI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054703.1 | XP_017910192.1 | 3 - 4 | 383 | 60 | |
| GenBank mRNA | KA337383.1 | - | 3 - 4 | 436 | 60 | |
More Information
Set Membership: |
|

PADI4
Genomic Map