Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046339.1 | XP_017901828.1 | 14 - 15 | 2198 | 61 | |
| XM_018046340.1 | XP_017901829.1 | 14 - 15 | 2149 | 61 | ||
| XM_018046341.1 | XP_017901830.1 | 13 - 14 | 2050 | 61 | ||
| XM_018046342.1 | XP_017901831.1 | 14 - 15 | 2868 | 61 | ||
| GenBank mRNA | JO369516.2 | - | 6 - 7 | 673 | 61 | |
More Information
Set Membership: |
|

KIRREL1
Genomic Map