Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046339.1 | XP_017901828.1 | 3 - 4 | 704 | 64 | |
| XM_018046340.1 | XP_017901829.1 | 3 - 4 | 703 | 64 | ||
| XM_018046342.1 | XP_017901831.1 | 3 - 4 | 1374 | 64 | ||
More Information
Set Membership: |
|

KIRREL1
Genomic Map