Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046339.1 | XP_017901828.1 | 7 - 8 | 1264 | 68 | |
| XM_018046340.1 | XP_017901829.1 | 7 - 8 | 1263 | 68 | ||
| XM_018046341.1 | XP_017901830.1 | 6 - 7 | 1116 | 68 | ||
| XM_018046342.1 | XP_017901831.1 | 7 - 8 | 1934 | 68 | ||
More Information
Set Membership: |
|

KIRREL1
Genomic Map