Assay Details
| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043802.1 | XP_017899291.1 | 5 - 6 | 648 | 62 | |
| XM_018043803.1 | XP_017899292.1 | 6 - 7 | 738 | 62 | ||
| XM_018043804.1 | XP_017899293.1 | 3 - 4 | 443 | 62 | ||
| XM_018043805.1 | XP_017899294.1 | 5 - 6 | 649 | 62 | ||
| XM_018043807.1 | XP_017899296.1 | 5 - 6 | 650 | 62 | ||
| GenBank mRNA | JO432196.2 | - | 5 - 6 | 824 | 62 | |
More Information
Set Membership: |
|

TAB3
Genomic Map