Assay Details
| Gene Symbol: S100A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977249.2 | XP_013832703.2 | 2 - 3 | 364 | 81 | |
| XM_018046132.1 | XP_017901621.1 | 2 - 3 | 293 | 81 | ||
More Information
Set Membership: |
|

S100A9
Genomic Map