Assay Details
| Gene Symbol: GPR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675569.3 | XP_005675626.2 | 2 - 3 | 1314 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR149| Gene Symbol: GPR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675569.3 | XP_005675626.2 | 2 - 3 | 1314 | 143 | |
Set Membership: |
|