Assay Details
Gene Symbol: LOC102186356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677150.3 | XP_005677207.2 | 4 - 5 | 758 | 87 | |
XM_005677151.3 | XP_005677208.2 | 4 - 5 | 758 | 87 | ||
XM_013975509.2 | XP_013830963.2 | 4 - 5 | 759 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
