Assay Details
| Gene Symbol: LOC102183200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046497.1 | XP_017901986.1 | 11 - 12 | 2168 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102183200| Gene Symbol: LOC102183200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046497.1 | XP_017901986.1 | 11 - 12 | 2168 | 84 | |
Set Membership: |
|