Assay Details
| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678658.3 | XP_005678715.1 | 5 - 6 | 884 | 57 | |
| XM_018042176.1 | XP_017897665.1 | 5 - 6 | 884 | 57 | ||
| GenBank mRNA | JO380396.2 | - | 1 - 2 | 535 | 57 | |
More Information
Set Membership: |
|

NT5C1A
Genomic Map