Assay Details
| Gene Symbol: MATR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683005.3 | XP_005683062.1 | 4 - 5 | 1331 | 66 | |
| XM_005683007.3 | XP_005683064.1 | 4 - 5 | 1357 | 66 | ||
| XM_005683008.3 | XP_005683065.1 | 4 - 5 | 1331 | 66 | ||
| XM_018050313.1 | XP_017905802.1 | 7 - 8 | 1504 | 66 | ||
| XM_018050314.1 | XP_017905803.1 | 4 - 5 | 1364 | 66 | ||
| XM_018050315.1 | XP_017905804.1 | 7 - 8 | 1502 | 66 | ||
| XM_018050316.1 | XP_017905805.1 | 4 - 5 | 1527 | 66 | ||
| XM_018050317.1 | XP_017905806.1 | 7 - 8 | 1504 | 66 | ||
| XM_018050318.1 | XP_017905807.1 | 7 - 8 | 1502 | 66 | ||
| XM_018050319.1 | XP_017905808.1 | 3 - 4 | 424 | 66 | ||
More Information
Set Membership: |
|

MATR3
Genomic Map