Assay Details
| Gene Symbol: DELE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683338.3 | XP_005683395.2 | 11 - 12 | 2227 | 86 | |
| XM_018050215.1 | XP_017905704.1 | 9 - 10 | 1975 | 86 | ||
More Information
Set Membership: |
|

DELE1
Genomic Map