Assay Details
| Gene Symbol: DELE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683338.3 | XP_005683395.2 | 6 - 7 | 1567 | 100 | |
| XM_018050215.1 | XP_017905704.1 | 6 - 7 | 1567 | 100 | ||
More Information
Set Membership: |
|

DELE1
Genomic Map