Assay Details
| Gene Symbol: ATG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683386.2 | XP_005683443.1 | 4 - 5 | 534 | 75 | |
| XM_005683388.3 | XP_005683445.1 | 4 - 5 | 534 | 75 | ||
| XM_005683389.3 | XP_005683446.1 | 4 - 5 | 534 | 75 | ||
More Information
Set Membership: |
|

ATG10
Genomic Map