Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684424.3 | XP_005684481.2 | 2 - 3 | 332 | 80 | |
| GenBank mRNA | KA339110.1 | - | 3 - 4 | 278 | 80 | |
More Information
Set Membership: |
|

ALKAL2
Genomic Map