Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684424.3 | XP_005684481.2 | 4 - 5 | 483 | 76 | |
| GenBank mRNA | KA339110.1 | - | 1 - 2 | 127 | 76 | |
More Information
Set Membership: |
|

ALKAL2
Genomic Map