Assay Details
| Gene Symbol: RNF219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687683.3 | XP_005687740.1 | 1 - 2 | 140 | 93 | |
| GenBank mRNA | JT705342.1 | - | 1 - 2 | 95 | 93 | |
More Information
Set Membership: |
|

RNF219
Genomic Map