Assay Details
| Gene Symbol: UPF3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044973.1 | XP_017900462.1 | 9 - 10 | 1366 | 69 | |
| GenBank mRNA | KA330779.1 | - | 1 - 2 | 836 | 69 | |
More Information
Set Membership: |
|

UPF3A
Genomic Map