Assay Details
| Gene Symbol: IMPAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688998.3 | XP_005689055.2 | 4 - 5 | 1069 | 71 | |
| GenBank mRNA | JT763441.1 | - | 1 - 2 | 633 | 71 | |
More Information
Set Membership: |
|

IMPAD1
Genomic Map