Assay Details
| Gene Symbol: KCNN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692619.3 | XP_005692676.2 | 7 - 8 | 1294 | 61 | |
| XM_013971419.2 | XP_013826873.2 | 7 - 8 | 1077 | 61 | ||
| GenBank mRNA | JO502686.2 | - | 5 - 6 | 500 | 61 | |
| JO509018.2 | - | 5 - 6 | 500 | 61 | ||
More Information
Set Membership: |
|

KCNN4
Genomic Map