Assay Details
| Gene Symbol: SHPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693410.3 | XP_005693467.2 | 5 - 6 | 1030 | 54 | |
| GenBank mRNA | KA346469.1 | - | 2 - 3 | 991 | 54 | |
More Information
Set Membership: |
|

SHPK
Genomic Map