Assay Details
| Gene Symbol: SNX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695119.3 | XP_005695176.1 | 1 - 2 | 1707 | 62 | |
| GenBank mRNA | JO425053.1 | - | 1 - 2 | 1726 | 62 | |
| KA317467.1 | - | 3134 | 62 | |||
More Information
Set Membership: |
|

SNX33
Genomic Map